Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LGR4
Entrez Gene ID
55366
Gene Name
leucine rich repeat containing G protein-coupled receptor 4
Gene Aliases
BNMD17, GPR48
Chromosome Location
Chr.11: 27365961 - 27472787 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018490.2 NP_060960.2 7 - 8 1239 139
GenBank mRNA AB209743.1 - 7 - 8 1158 139
AF257182.1 - 7 - 8 1198 139
BC033039.2 - 6 - 7 1155 139

View Full Product Details