The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015971.3 | NP_057055.2 | 2 - 3 | 507 | 87 |
GenBank mRNA | AF077042.1 | - | 2 - 3 | 456 | 87 |
AK222995.1 | - | 2 - 3 | 471 | 87 | |
AK313854.1 | - | 2 - 3 | 281 | 87 | |
BC000241.1 | - | 2 - 3 | 466 | 87 | |
BC050641.2 | - | 2 - 3 | 263 | 87 | |
BP379038.1 | - | 2 - 3 | 512 | 87 | |
BU599633.1 | - | 2 - 3 | 485 | 87 |