The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014001.4 | NP_054720.1 | 2 - 3 | 199 | 152 |
NM_138619.3 | NP_619525.1 | 2 - 3 | 199 | 152 | |
XM_011524563.2 | XP_011522865.1 | 2 - 3 | 173 | 152 | |
XM_017024385.1 | XP_016879874.1 | 2 - 3 | 173 | 152 | |
XM_017024386.1 | XP_016879875.1 | 2 - 3 | 161 | 152 | |
XM_017024391.1 | XP_016879880.1 | 2 - 3 | 156 | 152 | |
GenBank mRNA | AF190864.1 | - | 2 - 3 | 133 | 152 |
AF219138.1 | - | 2 - 3 | 134 | 152 | |
AF219139.1 | - | 2 - 3 | 134 | 152 | |
AK291651.1 | - | 2 - 3 | 164 | 152 | |
AK291685.1 | - | 2 - 3 | 130 | 152 | |
AK293291.1 | - | 2 - 3 | 164 | 152 | |
AK302423.1 | - | 2 - 3 | 128 | 152 | |
AK304692.1 | - | 2 - 3 | 128 | 152 | |
BC063285.1 | - | 2 - 3 | 161 | 152 | |
BC070044.1 | - | 2 - 3 | 138 | 152 | |
D63876.1 | - | 2 - 3 | 133 | 152 | |
DA312660.1 | - | 2 - 3 | 199 | 152 |