The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012761.2 | NP_001012779.2 | 1 - 2 | 328 | 56 |
XM_005271966.3 | XP_005272023.1 | 1 - 2 | 187 | 56 | |
GenBank mRNA | BC067736.1 | - | 1 - 2 | 328 | 56 |