The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000035.3 | NP_000026.2 | 7 - 8 | 884 | 67 |
GenBank mRNA | AK026411.1 | - | 7 - 8 | 661 | 67 |
AK290795.1 | - | 7 - 8 | 878 | 67 | |
BC029399.1 | - | 7 - 8 | 878 | 67 | |
X00270.1 | - | 1 - 2 | 91 | 67 | |
X01098.1 | - | 7 - 8 | 868 | 67 | |
X02747.1 | - | 7 - 8 | 927 | 67 |