The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001032296.3 | NP_001027467.2 | 7 - 8 | 1297 | 94 |
NM_001286649.1 | NP_001273578.1 | 6 - 7 | 1240 | 94 | |
NM_003576.4 | NP_003567.2 | 7 - 8 | 1241 | 94 | |
XM_005254079.4 | XP_005254136.1 | 7 - 8 | 1118 | 94 | |
XM_017020794.1 | XP_016876283.1 | 7 - 8 | 1414 | 94 | |
XM_017020795.1 | XP_016876284.1 | 7 - 8 | 1076 | 94 | |
GenBank mRNA | AF024636.1 | - | 7 - 8 | 1010 | 94 |
AF083420.1 | - | 7 - 8 | 1113 | 94 | |
AF339826.1 | - | 6 - 7 | 718 | 94 | |
AK297583.1 | - | 7 - 8 | 1013 | 94 | |
AK299140.1 | - | 6 - 7 | 957 | 94 | |
BC035578.1 | - | 7 - 8 | 1245 | 94 | |
BC065378.1 | - | 7 - 8 | 1241 | 94 | |
BT019619.1 | - | 7 - 8 | 932 | 94 | |
CR542227.1 | - | 7 - 8 | 932 | 94 | |
EU668350.1 | - | 7 - 8 | 1288 | 94 |