The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136035.2 | NP_001129507.1 | 5 - 6 | 770 | 64 |
NM_001142554.1 | NP_001136026.1 | 4 - 5 | 891 | 64 | |
NM_017722.3 | NP_060192.1 | 4 - 5 | 891 | 64 | |
XM_006722793.2 | XP_006722856.1 | 4 - 5 | 1013 | 64 | |
XM_011528124.1 | XP_011526426.1 | 5 - 6 | 689 | 64 | |
XM_011528125.1 | XP_011526427.1 | 6 - 7 | 1222 | 64 | |
XM_017026944.1 | XP_016882433.1 | 6 - 7 | 1213 | 64 | |
XM_017026945.1 | XP_016882434.1 | 6 - 7 | 1213 | 64 | |
XM_017026946.1 | XP_016882435.1 | 4 - 5 | 1013 | 64 | |
XM_017026947.1 | XP_016882436.1 | 6 - 7 | 1212 | 64 | |
GenBank mRNA | AF196479.1 | - | 4 - 5 | 641 | 64 |
AK000251.1 | - | 5 - 6 | 729 | 64 | |
AK292377.1 | - | 5 - 6 | 770 | 64 | |
AK295245.1 | - | 5 - 6 | 670 | 64 | |
AK304636.1 | - | 5 - 6 | 710 | 64 | |
BC002492.2 | - | 4 - 5 | 721 | 64 | |
BC018302.1 | - | 4 - 5 | 886 | 64 | |
BC040126.1 | - | 4 - 5 | 760 | 64 |