The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000131.4 | NP_000122.1 | 8 - 9 | 861 | 141 |
NM_001267554.1 | NP_001254483.1 | 5 - 6 | 609 | 141 | |
NM_019616.3 | NP_062562.1 | 7 - 8 | 795 | 141 | |
NR_051961.1 | 6 - 7 | 828 | 141 | ||
XM_006719963.3 | XP_006720026.2 | 6 - 7 | 669 | 141 | |
XM_011537474.2 | XP_011535776.2 | 8 - 9 | 918 | 141 | |
XM_011537475.2 | XP_011535777.2 | 6 - 7 | 732 | 141 | |
XM_011537476.2 | XP_011535778.1 | 5 - 6 | 1276 | 141 | |
GenBank mRNA | AK298404.1 | - | 5 - 6 | 606 | 141 |
AK311565.1 | - | 7 - 8 | 710 | 141 | |
AK311682.1 | - | 6 - 7 | 777 | 141 | |
BC130468.1 | - | 7 - 8 | 785 | 141 | |
EF421855.1 | - | 2 - 3 | 230 | 141 | |
EU557239.1 | - | 8 - 9 | 811 | 141 | |
M13232.1 | - | 8 - 9 | 842 | 141 |