The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139075.3 | NP_620714.2 | 8 - 9 | 947 | 60 |
XM_005273824.3 | XP_005273881.1 | 8 - 9 | 929 | 60 | |
XM_005273826.3 | XP_005273883.1 | 6 - 7 | 640 | 60 | |
XM_005273830.3 | XP_005273887.1 | 7 - 8 | 771 | 60 | |
XM_005273831.3 | XP_005273888.1 | 5 - 6 | 623 | 60 | |
XM_011544802.2 | XP_011543104.1 | 7 - 8 | 820 | 60 | |
XM_011544805.1 | XP_011543107.1 | 8 - 9 | 932 | 60 | |
XM_011544806.1 | XP_011543108.1 | 8 - 9 | 932 | 60 | |
XM_011544807.2 | XP_011543109.1 | 7 - 8 | 768 | 60 | |
XM_017017328.1 | XP_016872817.1 | 8 - 9 | 932 | 60 | |
XM_017017329.1 | XP_016872818.1 | 8 - 9 | 929 | 60 | |
GenBank mRNA | AB209253.1 | - | 5 - 6 | 593 | 60 |
AK123089.1 | - | 6 - 7 | 640 | 60 | |
AK298919.1 | - | 8 - 9 | 868 | 60 | |
AY029200.1 | - | 8 - 9 | 932 | 60 | |
BC063008.1 | - | 8 - 9 | 883 | 60 | |
CD636711.1 | - | 5 - 6 | 60 |