The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000693.3 | NP_000684.2 | 4 - 5 | 696 | 63 |
GenBank mRNA | AK312303.1 | - | 4 - 5 | 557 | 63 |
BC069274.1 | - | 4 - 5 | 536 | 63 | |
U07919.1 | - | 4 - 5 | 63 |