The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080496.2 | NP_001073965.2 | 3 - 4 | 401 | 71 |
GenBank mRNA | AK289566.1 | - | 3 - 4 | 394 | 71 |
BC001725.1 | - | 3 - 4 | 329 | 71 | |
D87447.1 | - | 3 - 4 | 338 | 71 |