The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032117.3 | NP_115493.1 | 6 - 7 | 554 | 107 |
NR_045605.1 | 8 - 9 | 715 | 107 | ||
XM_017008688.1 | XP_016864177.1 | 6 - 7 | 911 | 107 | |
GenBank mRNA | AK312172.1 | - | 6 - 7 | 513 | 107 |
AY028916.1 | - | 6 - 7 | 493 | 107 | |
BC032142.2 | - | 6 - 7 | 508 | 107 | |
BG758047.1 | - | 8 - 9 | 684 | 107 | |
BU533354.1 | - | 3 - 4 | 210 | 107 |