Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRMT7
Entrez Gene ID
54496
Gene Name
protein arginine methyltransferase 7
Gene Aliases
-
Chromosome Location
Chr.16: 68310974 - 68358832 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290018.1 NP_001276947.1 3 - 4 941 111
NM_019023.2 NP_061896.1 3 - 4 435 111
XM_011523112.2 XP_011521414.1 2 - 3 300 111
XM_011523113.2 XP_011521415.1 3 - 4 427 111
XM_011523115.2 XP_011521417.1 2 - 3 244 111
XM_011523116.2 XP_011521418.1 2 - 3 300 111
XM_011523128.2 XP_011521430.1 3 - 4 353 111
XM_017023290.1 XP_016878779.1 3 - 4 427 111
XM_017023291.1 XP_016878780.1 3 - 4 367 111
XM_017023292.1 XP_016878781.1 2 - 3 244 111
XM_017023293.1 XP_016878782.1 3 - 4 427 111
XM_017023294.1 XP_016878783.1 3 - 4 367 111
XM_017023295.1 XP_016878784.1 2 - 3 300 111
XM_017023296.1 XP_016878785.1 3 - 4 427 111
XM_017023297.1 XP_016878786.1 3 - 4 427 111
XM_017023298.1 XP_016878787.1 3 - 4 427 111
XM_017023299.1 XP_016878788.1 3 - 4 427 111
XM_017023300.1 XP_016878789.1 3 - 4 427 111
XM_017023301.1 XP_016878790.1 3 - 4 427 111
XM_017023304.1 XP_016878793.1 3 - 4 427 111
XM_017023306.1 XP_016878795.1 3 - 4 427 111
XM_017023311.1 XP_016878800.1 2 - 3 260 111
XM_017023312.1 XP_016878801.1 2 - 3 300 111
XM_017023314.1 XP_016878803.1 2 - 3 211 111
XM_017023316.1 XP_016878805.1 2 - 3 211 111
GenBank mRNA AK001502.1 - 3 - 4 367 111
AK056647.1 - 3 - 4 923 111
AK097175.1 - 1 - 2 877 111
BC000146.1 - 1 - 2 176 111
DA287041.1 - 3 - 4 414 111
DA648148.1 - 2 - 3 300 111

View Full Product Details