Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRMT7
Entrez Gene ID
54496
Gene Name
protein arginine methyltransferase 7
Gene Aliases
-
Chromosome Location
Chr.16: 68310974 - 68358832 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184824.1 NP_001171753.1 6 - 7 804 69
NM_001290018.1 NP_001276947.1 8 - 9 1595 69
NM_019023.2 NP_061896.1 8 - 9 1089 69
XM_011523112.2 XP_011521414.1 7 - 8 954 69
XM_011523113.2 XP_011521415.1 8 - 9 1081 69
XM_011523115.2 XP_011521417.1 7 - 8 898 69
XM_011523116.2 XP_011521418.1 7 - 8 954 69
XM_011523121.2 XP_011521423.1 5 - 6 719 69
XM_011523124.2 XP_011521426.1 7 - 8 1007 69
XM_011523125.2 XP_011521427.1 6 - 7 849 69
XM_011523126.2 XP_011521428.1 6 - 7 853 69
XM_011523128.2 XP_011521430.1 7 - 8 898 69
XM_011523131.2 XP_011521433.1 6 - 7 828 69
XM_017023290.1 XP_016878779.1 8 - 9 1081 69
XM_017023291.1 XP_016878780.1 8 - 9 1021 69
XM_017023292.1 XP_016878781.1 7 - 8 898 69
XM_017023293.1 XP_016878782.1 8 - 9 1081 69
XM_017023294.1 XP_016878783.1 8 - 9 1021 69
XM_017023295.1 XP_016878784.1 7 - 8 954 69
XM_017023296.1 XP_016878785.1 8 - 9 1081 69
XM_017023297.1 XP_016878786.1 8 - 9 1081 69
XM_017023298.1 XP_016878787.1 8 - 9 1081 69
XM_017023299.1 XP_016878788.1 8 - 9 1081 69
XM_017023300.1 XP_016878789.1 8 - 9 1081 69
XM_017023301.1 XP_016878790.1 8 - 9 1081 69
XM_017023302.1 XP_016878791.1 6 - 7 849 69
XM_017023303.1 XP_016878792.1 7 - 8 1007 69
XM_017023304.1 XP_016878793.1 8 - 9 1081 69
XM_017023305.1 XP_016878794.1 5 - 6 719 69
XM_017023306.1 XP_016878795.1 8 - 9 1081 69
XM_017023307.1 XP_016878796.1 7 - 8 1007 69
XM_017023308.1 XP_016878797.1 7 - 8 1021 69
XM_017023309.1 XP_016878798.1 6 - 7 849 69
XM_017023310.1 XP_016878799.1 6 - 7 873 69
XM_017023311.1 XP_016878800.1 6 - 7 805 69
XM_017023312.1 XP_016878801.1 7 - 8 954 69
XM_017023313.1 XP_016878802.1 7 - 8 996 69
XM_017023314.1 XP_016878803.1 6 - 7 756 69
XM_017023315.1 XP_016878804.1 7 - 8 996 69
XM_017023316.1 XP_016878805.1 6 - 7 756 69
GenBank mRNA AB067520.1 - 2 - 3 1091 69
AK001502.1 - 8 - 9 1021 69
AK022739.1 - 4 - 5 516 69
AK097175.1 - 6 - 7 69
AK304605.1 - 6 - 7 727 69
BC000146.1 - 6 - 7 830 69

View Full Product Details