Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SYNRG
- Entrez Gene ID
-
11276
- Gene Name
- synergin, gamma
- Gene Aliases
-
AP1GBP1, SYNG
- Chromosome Location
-
Chr.17: 37514797 - 37613637 on Build GRCh38
View Full Product Details