The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003559.4 | NP_003550.1 | 9 - 10 | 1652 | 63 |
XM_011525326.2 | XP_011523628.1 | 10 - 11 | 1793 | 63 | |
XM_011525327.2 | XP_011523629.1 | 11 - 12 | 1312 | 63 | |
XM_011525330.2 | XP_011523632.1 | 8 - 9 | 988 | 63 | |
XM_017025196.1 | XP_016880685.1 | 10 - 11 | 1229 | 63 | |
XM_017025197.1 | XP_016880686.1 | 10 - 11 | 1129 | 63 | |
XM_017025198.1 | XP_016880687.1 | 8 - 9 | 1615 | 63 | |
XM_017025199.1 | XP_016880688.1 | 9 - 10 | 1067 | 63 | |
GenBank mRNA | BT019614.1 | - | 9 - 10 | 1171 | 63 |
U85245.1 | - | 9 - 10 | 1651 | 63 |