Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KIAA0226L
Entrez Gene ID
80183
Gene Name
KIAA0226 like
Gene Aliases
C13orf18
Chromosome Location
Chr.13: 46342000 - 46390042 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286761.1 NP_001273690.1 6 - 7 1508 86
NM_001286762.1 NP_001273691.1 5 - 6 1414 86
NM_001286763.1 NP_001273692.1 6 - 7 1143 86
NM_001286764.1 NP_001273693.1 5 - 6 1027 86
NM_001286765.1 NP_001273694.1 5 - 6 873 86
NM_001286766.1 NP_001273695.1 4 - 5 757 86
NM_025113.3 NP_079389.2 6 - 7 1530 86
GenBank mRNA AK025215.1 - 5 - 6 1163 86
AK093073.1 - 6 - 7 1478 86
AK125950.1 - 6 - 7 1515 86
AK293034.1 - 5 - 6 779 86
AK300183.1 - 5 - 6 577 86
AK307854.1 - 4 - 5 500 86
AK310264.1 - 7 - 8 1517 86
BC004495.1 - 2 - 3 227 86
BC043488.1 - 6 - 7 876 86
BX641126.1 - 4 - 5 564 86

View Full Product Details