The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286761.1 | NP_001273690.1 | 7 - 8 | 1560 | 157 |
NM_001286762.1 | NP_001273691.1 | 6 - 7 | 1466 | 157 | |
NM_001286763.1 | NP_001273692.1 | 7 - 8 | 1195 | 157 | |
NM_001286764.1 | NP_001273693.1 | 6 - 7 | 1079 | 157 | |
NM_001286765.1 | NP_001273694.1 | 6 - 7 | 925 | 157 | |
NM_001286766.1 | NP_001273695.1 | 5 - 6 | 809 | 157 | |
NM_025113.3 | NP_079389.2 | 7 - 8 | 1582 | 157 | |
GenBank mRNA | AK025215.1 | - | 6 - 7 | 1215 | 157 |
AK093073.1 | - | 7 - 8 | 1530 | 157 | |
AK293034.1 | - | 6 - 7 | 831 | 157 | |
AK300183.1 | - | 6 - 7 | 629 | 157 | |
AK307854.1 | - | 5 - 6 | 552 | 157 | |
AK310264.1 | - | 8 - 9 | 1569 | 157 | |
BC004495.1 | - | 3 - 4 | 279 | 157 | |
BC043488.1 | - | 7 - 8 | 928 | 157 | |
BX641126.1 | - | 5 - 6 | 616 | 157 |