The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004589.3 | NP_004580.1 | 4 - 5 | 723 | 77 |
XM_005256751.3 | XP_005256808.1 | 4 - 5 | 690 | 77 | |
GenBank mRNA | AF026852.1 | - | 4 - 5 | 675 | 77 |
AF131816.1 | - | 3 - 4 | 330 | 77 | |
AF183424.1 | - | 4 - 5 | 675 | 77 | |
AK315595.1 | - | 4 - 5 | 701 | 77 | |
BC015504.2 | - | 4 - 5 | 678 | 77 |