Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
APP
Entrez Gene ID
351
Gene Name
amyloid beta precursor protein
Gene Aliases
AAA, ABETA, ABPP, AD1, APPI, CTFgamma, CVAP, PN-II, PN2
Chromosome Location
Chr.21: 25880550 - 26171128 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000484.3 NP_000475.1 16 - 17 2266 75
NM_001136016.3 NP_001129488.1 15 - 16 2190 75
NM_001136129.2 NP_001129601.1 13 - 14 1873 75
NM_001136130.2 NP_001129602.1 15 - 16 2098 75
NM_001136131.2 NP_001129603.1 14 - 15 1928 75
NM_001204301.1 NP_001191230.1 15 - 16 2212 75
NM_001204302.1 NP_001191231.1 14 - 15 2155 75
NM_001204303.1 NP_001191232.1 13 - 14 1987 75
NM_201413.2 NP_958816.1 15 - 16 2209 75
NM_201414.2 NP_958817.1 14 - 15 2041 75
GenBank mRNA A02759.1 - 14 - 15 1987 75
AF282245.1 - 13 - 14 1800 75
AK294534.1 - 15 - 16 2045 75
AK295373.1 - 6 - 7 766 75
AK295621.1 - 15 - 16 2151 75
AK297229.1 - 12 - 13 1404 75
AK297412.1 - 13 - 14 1770 75
AK298861.1 - 13 - 14 1943 75
AK312326.1 - 16 - 17 2131 75
BC065529.1 - 15 - 16 2140 75
M15532.1 - - 47 75
M15533.1 - 1 - 2 47 75
M16765.1 - 9 - 10 1001 75
M18734.1 - 5 - 6 549 75
S60721.1 - 1 - 2 104 75
S61380.1 - 1 - 2 104 75
S61383.1 - 1 - 2 104 75
X06989.1 - 15 - 16 2133 75
Y00264.1 - 14 - 15 1988 75

View Full Product Details