The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000484.3 | NP_000475.1 | 17 - 18 | 2406 | 152 |
NM_001136016.3 | NP_001129488.1 | 16 - 17 | 2330 | 152 | |
NM_001136129.2 | NP_001129601.1 | 14 - 15 | 2013 | 152 | |
NM_001136130.2 | NP_001129602.1 | 16 - 17 | 2238 | 152 | |
NM_001136131.2 | NP_001129603.1 | 15 - 16 | 2068 | 152 | |
NM_001204301.1 | NP_001191230.1 | 16 - 17 | 2352 | 152 | |
NM_001204302.1 | NP_001191231.1 | 15 - 16 | 2295 | 152 | |
NM_001204303.1 | NP_001191232.1 | 14 - 15 | 2127 | 152 | |
NM_201413.2 | NP_958816.1 | 16 - 17 | 2349 | 152 | |
NM_201414.2 | NP_958817.1 | 15 - 16 | 2181 | 152 | |
GenBank mRNA | A02759.1 | - | 15 - 16 | 2127 | 152 |
AF282245.1 | - | 14 - 15 | 1940 | 152 | |
AK294534.1 | - | 16 - 17 | 2185 | 152 | |
AK295373.1 | - | 7 - 8 | 906 | 152 | |
AK295621.1 | - | 16 - 17 | 2291 | 152 | |
AK296229.1 | - | 15 - 16 | 2068 | 152 | |
AK297229.1 | - | 13 - 14 | 1544 | 152 | |
AK297412.1 | - | 14 - 15 | 1910 | 152 | |
AK298861.1 | - | 14 - 15 | 2083 | 152 | |
AK312326.1 | - | 17 - 18 | 2271 | 152 | |
BC065529.1 | - | 16 - 17 | 2280 | 152 | |
M15533.1 | - | 2 - 3 | 187 | 152 | |
M16765.1 | - | 10 - 11 | 1141 | 152 | |
M18734.1 | - | 6 - 7 | 689 | 152 | |
X06989.1 | - | 16 - 17 | 2273 | 152 | |
Y00264.1 | - | 15 - 16 | 2128 | 152 |