Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
APP
Entrez Gene ID
351
Gene Name
amyloid beta precursor protein
Gene Aliases
AAA, ABETA, ABPP, AD1, APPI, CTFgamma, CVAP, PN-II, PN2
Chromosome Location
Chr.21: 25880550 - 26171128 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000484.3 NP_000475.1 9 - 10 1424 87
NM_001136016.3 NP_001129488.1 8 - 9 1348 87
NM_001136129.2 NP_001129601.1 6 - 7 1031 87
NM_001136130.2 NP_001129602.1 8 - 9 1256 87
NM_001136131.2 NP_001129603.1 7 - 8 1086 87
NM_001204301.1 NP_001191230.1 9 - 10 1424 87
NM_001204302.1 NP_001191231.1 8 - 9 1367 87
NM_001204303.1 NP_001191232.1 7 - 8 1199 87
NM_201413.2 NP_958816.1 8 - 9 1367 87
NM_201414.2 NP_958817.1 7 - 8 1199 87
GenBank mRNA A02759.1 - 7 - 8 1145 87
AF282245.1 - 6 - 7 958 87
AK294534.1 - 8 - 9 1203 87
AK295621.1 - 8 - 9 1309 87
AK296229.1 - 7 - 8 1086 87
AK297229.1 - 5 - 6 562 87
AK297412.1 - 6 - 7 928 87
AK298861.1 - 6 - 7 1101 87
AK312326.1 - 9 - 10 1289 87
BC065529.1 - 8 - 9 1298 87
M16765.1 - 2 - 3 159 87
M28373.1 - 6 - 7 930 87
X06989.1 - 8 - 9 1291 87
Y00264.1 - 7 - 8 1146 87

View Full Product Details