The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025185.3 | NP_079461.2 | 22 - 23 | 3778 | 69 |
XM_005257203.4 | XP_005257260.1 | 23 - 24 | 4005 | 69 | |
XM_006721806.3 | XP_006721869.1 | 25 - 26 | 4257 | 69 | |
XM_006721807.3 | XP_006721870.1 | 24 - 25 | 4227 | 69 | |
XM_006721810.3 | XP_006721873.1 | 25 - 26 | 4257 | 69 | |
XM_006721811.3 | XP_006721874.1 | 24 - 25 | 4227 | 69 | |
XM_011524597.2 | XP_011522899.1 | 25 - 26 | 4257 | 69 | |
XM_011524598.2 | XP_011522900.1 | 24 - 25 | 4227 | 69 | |
XM_011524599.1 | XP_011522901.1 | 25 - 26 | 4083 | 69 | |
XM_011524600.2 | XP_011522902.1 | 24 - 25 | 4146 | 69 | |
XM_011524601.2 | XP_011522903.1 | 25 - 26 | 4146 | 69 | |
XM_011524602.2 | XP_011522904.1 | 24 - 25 | 4035 | 69 | |
XM_017024429.1 | XP_016879918.1 | 24 - 25 | 4053 | 69 | |
XM_017024430.1 | XP_016879919.1 | 21 - 22 | 7424 | 69 | |
GenBank mRNA | AB046856.2 | - | 18 - 19 | 3379 | 69 |