The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025185.3 | NP_079461.2 | 23 - 24 | 3940 | 64 |
XM_005257203.4 | XP_005257260.1 | 24 - 25 | 4167 | 64 | |
XM_006721806.3 | XP_006721869.1 | 26 - 27 | 4419 | 64 | |
XM_006721807.3 | XP_006721870.1 | 25 - 26 | 4389 | 64 | |
XM_011524597.2 | XP_011522899.1 | 26 - 27 | 4419 | 64 | |
XM_011524598.2 | XP_011522900.1 | 25 - 26 | 4389 | 64 | |
XM_011524599.1 | XP_011522901.1 | 26 - 27 | 4245 | 64 | |
XM_011524600.2 | XP_011522902.1 | 25 - 26 | 4308 | 64 | |
XM_011524601.2 | XP_011522903.1 | 26 - 27 | 4308 | 64 | |
XM_011524602.2 | XP_011522904.1 | 25 - 26 | 4197 | 64 | |
XM_017024429.1 | XP_016879918.1 | 25 - 26 | 4215 | 64 | |
XM_017024430.1 | XP_016879919.1 | 22 - 23 | 7586 | 64 | |
GenBank mRNA | AB046856.2 | - | 19 - 20 | 3541 | 64 |
AJ278120.1 | - | 10 - 11 | 1344 | 64 |