Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FIBCD1
Entrez Gene ID
84929
Gene Name
fibrinogen C domain containing 1
Gene Aliases
-
Chromosome Location
Chr.9: 130902438 - 130939068 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145106.1 NP_001138578.1 5 - 6 961 93
NM_032843.4 NP_116232.3 4 - 5 1099 93
GenBank mRNA AK027716.1 - 3 - 4 715 93
AK292489.1 - 4 - 5 868 93
AY358311.1 - 4 - 5 856 93
BC032953.2 - 4 - 5 1097 93
BC112892.1 - 4 - 5 93

View Full Product Details