Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RTTN
Entrez Gene ID
25914
Gene Name
rotatin
Gene Aliases
MSSP
Chromosome Location
Chr.18: 70003804 - 70205726 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318520.1 NP_001305449.1 34 - 35 4634 58
NM_173630.3 NP_775901.3 35 - 36 4817 58
XM_006722434.3 XP_006722497.1 35 - 36 4820 58
XM_006722435.3 XP_006722498.1 35 - 36 4820 58
XM_011525902.2 XP_011524204.1 34 - 35 4580 58
XM_011525903.2 XP_011524205.1 33 - 34 4391 58
XM_011525904.2 XP_011524206.1 35 - 36 4820 58
XM_011525905.2 XP_011524207.1 35 - 36 4820 58
XM_011525906.2 XP_011524208.1 25 - 26 4101 58
XM_017025693.1 XP_016881182.1 34 - 35 4577 58
XM_017025694.1 XP_016881183.1 32 - 33 4375 58
XM_017025695.1 XP_016881184.1 28 - 29 3877 58
XM_017025696.1 XP_016881185.1 35 - 36 4817 58
GenBank mRNA BC047602.1 - 27 - 28 3725 58
BC143723.1 - 34 - 35 4611 58

View Full Product Details