Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PCK1
Entrez Gene ID
5105
Gene Name
phosphoenolpyruvate carboxykinase 1
Gene Aliases
PEPCK-C, PEPCK1, PEPCKC
Chromosome Location
Chr.20: 57561081 - 57566461 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002591.3 NP_002582.3 6 - 7 1122 86
XM_011528839.1 XP_011527141.1 4 - 5 764 86
GenBank mRNA AK290802.1 - 6 - 7 1100 86
AK303806.1 - 4 - 5 674 86
BC023978.1 - 6 - 7 1097 86
L05144.1 - 6 - 7 1080 86

View Full Product Details