Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PCK1
Entrez Gene ID
5105
Gene Name
phosphoenolpyruvate carboxykinase 1
Gene Aliases
PEPCK-C, PEPCK1, PEPCKC
Chromosome Location
Chr.20: 57561081 - 57566461 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002591.3 NP_002582.3 7 - 8 1354 81
XM_011528839.1 XP_011527141.1 5 - 6 996 81
GenBank mRNA AK290802.1 - 7 - 8 1332 81
AK300072.1 - 3 - 4 588 81
BC023978.1 - 7 - 8 1329 81
L05144.1 - 7 - 8 1312 81

View Full Product Details