Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CLEC2B
- Entrez Gene ID
-
9976
- Gene Name
- C-type lectin domain family 2 member B
- Gene Aliases
-
AICL, CLECSF2, HP10085, IFNRG1
- Chromosome Location
-
Chr.12: 9852369 - 9869859 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK311232.1 |
- |
1 - 2 |
146 |
76 |
View Full Product Details