The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278463.1 | NP_001265392.1 | 17 - 18 | 2128 | 142 |
NM_001278464.1 | NP_001265393.1 | 19 - 20 | 2200 | 142 | |
NM_001278465.1 | NP_001265394.1 | 18 - 19 | 2167 | 142 | |
NM_001278466.1 | NP_001265395.1 | 15 - 16 | 1757 | 142 | |
NM_005690.4 | NP_005681.2 | 16 - 17 | 2050 | 142 | |
NM_012062.4 | NP_036192.2 | 18 - 19 | 2161 | 142 | |
NM_012063.3 | NP_036193.2 | 17 - 18 | 2083 | 142 | |
XM_005253282.4 | XP_005253339.1 | 17 - 18 | 2061 | 142 | |
XM_011520543.2 | XP_011518845.1 | 18 - 19 | 2094 | 142 | |
XM_011520544.2 | XP_011518846.1 | 17 - 18 | 1878 | 142 | |
XM_017018663.1 | XP_016874152.1 | 16 - 17 | 1845 | 142 | |
XM_017018664.1 | XP_016874153.1 | 16 - 17 | 1800 | 142 | |
XM_017018665.1 | XP_016874154.1 | 15 - 16 | 1767 | 142 | |
GenBank mRNA | AB006965.1 | - | 18 - 19 | 2032 | 142 |
AB209070.1 | - | 18 - 19 | 2044 | 142 | |
AF000430.1 | - | 18 - 19 | 2069 | 142 | |
AF061795.1 | - | 16 - 17 | 1907 | 142 | |
AF151685.1 | - | 17 - 18 | 1985 | 142 | |
AK291094.1 | - | 16 - 17 | 1930 | 142 | |
AK294533.1 | - | 15 - 16 | 1637 | 142 | |
AK299926.1 | - | 19 - 20 | 2113 | 142 | |
AK302565.1 | - | 17 - 18 | 2083 | 142 | |
BC024590.1 | - | 17 - 18 | 1953 | 142 |