The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278463.1 | NP_001265392.1 | 1 - 2 | 263 | 69 |
NM_001278464.1 | NP_001265393.1 | 1 - 2 | 263 | 69 | |
NM_001278465.1 | NP_001265394.1 | 1 - 2 | 263 | 69 | |
NM_001278466.1 | NP_001265395.1 | 1 - 2 | 263 | 69 | |
NM_005690.4 | NP_005681.2 | 1 - 2 | 263 | 69 | |
NM_012062.4 | NP_036192.2 | 1 - 2 | 263 | 69 | |
NM_012063.3 | NP_036193.2 | 1 - 2 | 263 | 69 | |
XM_005253282.4 | XP_005253339.1 | 1 - 2 | 235 | 69 | |
XM_011520543.2 | XP_011518845.1 | 1 - 2 | 235 | 69 | |
XM_011520544.2 | XP_011518846.1 | 1 - 2 | 143 | 69 | |
XM_017018663.1 | XP_016874152.1 | 1 - 2 | 143 | 69 | |
XM_017018664.1 | XP_016874153.1 | 1 - 2 | 143 | 69 | |
XM_017018665.1 | XP_016874154.1 | 1 - 2 | 143 | 69 | |
GenBank mRNA | AB006965.1 | - | 1 - 2 | 134 | 69 |
AB209070.1 | - | 1 - 2 | 140 | 69 | |
AF000430.1 | - | 1 - 2 | 171 | 69 | |
AF061795.1 | - | 1 - 2 | 120 | 69 | |
AF151685.1 | - | 1 - 2 | 120 | 69 | |
AK291094.1 | - | 1 - 2 | 143 | 69 | |
AK293290.1 | - | 1 - 2 | 143 | 69 | |
AK294533.1 | - | 1 - 2 | 143 | 69 | |
AK298568.1 | - | 1 - 2 | 143 | 69 | |
AK299926.1 | - | 1 - 2 | 176 | 69 | |
AK302565.1 | - | 1 - 2 | 263 | 69 | |
AK311606.1 | - | 1 - 2 | 258 | 69 | |
BC024590.1 | - | 1 - 2 | 133 | 69 | |
BC107763.1 | - | 1 - 2 | 147 | 69 | |
DC377686.1 | - | 1 - 2 | 260 | 69 |