The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278463.1 | NP_001265392.1 | 9 - 10 | 1246 | 155 |
NM_001278464.1 | NP_001265393.1 | 10 - 11 | 1285 | 155 | |
NM_001278465.1 | NP_001265394.1 | 10 - 11 | 1285 | 155 | |
NM_001278466.1 | NP_001265395.1 | 6 - 7 | 842 | 155 | |
NM_005690.4 | NP_005681.2 | 9 - 10 | 1246 | 155 | |
NM_012062.4 | NP_036192.2 | 9 - 10 | 1246 | 155 | |
NM_012063.3 | NP_036193.2 | 9 - 10 | 1246 | 155 | |
XM_005253282.4 | XP_005253339.1 | 10 - 11 | 1257 | 155 | |
XM_011520543.2 | XP_011518845.1 | 10 - 11 | 1257 | 155 | |
XM_011520544.2 | XP_011518846.1 | 8 - 9 | 963 | 155 | |
XM_017018663.1 | XP_016874152.1 | 8 - 9 | 963 | 155 | |
XM_017018664.1 | XP_016874153.1 | 8 - 9 | 963 | 155 | |
XM_017018665.1 | XP_016874154.1 | 8 - 9 | 963 | 155 | |
GenBank mRNA | AB006965.1 | - | 9 - 10 | 1117 | 155 |
AB209070.1 | - | 10 - 11 | 1162 | 155 | |
AF000430.1 | - | 9 - 10 | 1154 | 155 | |
AF061795.1 | - | 9 - 10 | 1103 | 155 | |
AF151685.1 | - | 9 - 10 | 1103 | 155 | |
AK291094.1 | - | 9 - 10 | 1126 | 155 | |
AK293290.1 | - | 7 - 8 | 918 | 155 | |
AK294533.1 | - | 6 - 7 | 722 | 155 | |
AK298568.1 | - | 8 - 9 | 963 | 155 | |
AK299926.1 | - | 10 - 11 | 1198 | 155 | |
AK302565.1 | - | 9 - 10 | 1246 | 155 | |
AK311606.1 | - | 9 - 10 | 1241 | 155 | |
BC024590.1 | - | 9 - 10 | 1116 | 155 | |
BC107763.1 | - | 9 - 10 | 1130 | 155 |