The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278464.1 | NP_001265393.1 | 16 - 17 | 1872 | 147 |
NM_001278466.1 | NP_001265395.1 | 12 - 13 | 1429 | 147 | |
NM_012062.4 | NP_036192.2 | 15 - 16 | 1833 | 147 | |
XM_011520544.2 | XP_011518846.1 | 14 - 15 | 1550 | 147 | |
GenBank mRNA | AB006965.1 | - | 15 - 16 | 1704 | 147 |
AF000430.1 | - | 15 - 16 | 1741 | 147 | |
AK294533.1 | - | 12 - 13 | 1309 | 147 | |
AK299926.1 | - | 16 - 17 | 1785 | 147 |