The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318510.1 | NP_001305439.1 | 1 - 2 | 270 | 136 |
NM_004458.2 | NP_004449.1 | 1 - 2 | 259 | 133 | |
GenBank mRNA | AB061713.1 | - | 1 - 2 | 150 | 136 |
AB061714.1 | - | 1 - 2 | 110 | 136 | |
AF030555.1 | - | 1 - 2 | 260 | 133 | |
AK292070.1 | - | 1 - 2 | 152 | 136 | |
BC034959.2 | - | 1 - 2 | 122 | 133 | |
DR004263.1 | - | 1 - 2 | 277 | 136 | |
Y12777.1 | - | 1 - 2 | 120 | 136 |