The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001382.3 | NP_001373.2 | 6 - 7 | 1338 | 81 |
XM_005271422.2 | XP_005271479.1 | 6 - 7 | 1354 | 81 | |
XM_011542648.1 | XP_011540950.1 | 5 - 6 | 1556 | 81 | |
XM_017017293.1 | XP_016872782.1 | 5 - 6 | 1556 | 81 | |
XM_017017295.1 | XP_016872784.1 | 4 - 5 | 785 | 81 | |
GenBank mRNA | AK095718.1 | - | 4 - 5 | 1547 | 81 |
AK128572.1 | - | 8 - 9 | 2470 | 81 | |
AK225994.1 | - | 6 - 7 | 1083 | 81 | |
AK301973.1 | - | 5 - 6 | 1020 | 81 | |
AK312783.1 | - | 6 - 7 | 1141 | 81 | |
BC000325.2 | - | 6 - 7 | 1086 | 81 | |
BC047771.1 | - | 5 - 6 | 967 | 81 | |
BT006802.1 | - | 6 - 7 | 918 | 81 | |
Z82022.1 | - | 6 - 7 | 997 | 81 |