The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005253154.4 | XP_005253211.1 | 29 - 30 | 4042 | 85 |
XM_005253155.4 | XP_005253212.1 | 28 - 29 | 3910 | 85 | |
XM_011520394.2 | XP_011518696.1 | 28 - 29 | 3925 | 85 | |
XM_017018372.1 | XP_016873861.1 | 28 - 29 | 5331 | 85 | |
XM_017018373.1 | XP_016873862.1 | 29 - 30 | 3961 | 85 |