The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001725.2 | NP_001716.2 | 1 - 2 | 203 | 84 |
GenBank mRNA | AF322588.1 | - | 1 - 2 | 129 | 84 |
AK315328.1 | - | 1 - 2 | 170 | 84 | |
AX713278.1 | - | 1 - 2 | 171 | 84 | |
AX746123.1 | - | 1 - 2 | 171 | 84 | |
AX752831.1 | - | 1 - 2 | 171 | 84 | |
BC040955.1 | - | 1 - 2 | 178 | 84 | |
BP306215.1 | - | 1 - 2 | 215 | 84 | |
DQ414688.1 | - | 1 - 2 | 141 | 84 | |
J04739.1 | - | 1 - 2 | 171 | 84 |