The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318253.1 | NP_001305182.1 | 11 - 12 | 1505 | 143 |
NM_133171.4 | NP_573403.1 | 11 - 12 | 1437 | 143 | |
NM_182764.2 | NP_877496.1 | 10 - 11 | 1333 | 143 | |
XM_005260496.3 | XP_005260553.1 | 11 - 12 | 1002 | 143 | |
XM_005260498.3 | XP_005260555.1 | 10 - 11 | 1364 | 143 | |
XM_005260499.3 | XP_005260556.1 | 10 - 11 | 1335 | 143 | |
XM_005260500.3 | XP_005260557.1 | 9 - 10 | 863 | 143 | |
XM_005260501.4 | XP_005260558.1 | 10 - 11 | 873 | 143 | |
XM_006723854.3 | XP_006723917.1 | 10 - 11 | 936 | 143 | |
XM_017028009.1 | XP_016883498.1 | 10 - 11 | 1308 | 143 | |
XM_017028010.1 | XP_016883499.1 | 9 - 10 | 811 | 143 | |
XM_017028011.1 | XP_016883500.1 | 9 - 10 | 1264 | 143 | |
GenBank mRNA | AB058737.1 | - | 10 - 11 | 948 | 143 |
AF398886.1 | - | 9 - 10 | 876 | 143 | |
AF417861.1 | - | 9 - 10 | 808 | 143 | |
AK057032.1 | - | 11 - 12 | 961 | 143 | |
AK293861.1 | - | 9 - 10 | 901 | 143 | |
AK299318.1 | - | 13 - 14 | 1048 | 143 | |
BC000143.4 | - | 10 - 11 | 1013 | 143 | |
BX648819.1 | - | 10 - 11 | 914 | 143 |