The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318253.1 | NP_001305182.1 | 12 - 13 | 1623 | 86 |
NM_133171.4 | NP_573403.1 | 12 - 13 | 1555 | 86 | |
NM_182764.2 | NP_877496.1 | 11 - 12 | 1451 | 86 | |
XM_005260496.3 | XP_005260553.1 | 12 - 13 | 1120 | 86 | |
XM_005260498.3 | XP_005260555.1 | 11 - 12 | 1482 | 86 | |
XM_005260499.3 | XP_005260556.1 | 11 - 12 | 1453 | 86 | |
XM_005260500.3 | XP_005260557.1 | 10 - 11 | 981 | 86 | |
XM_005260501.4 | XP_005260558.1 | 11 - 12 | 991 | 86 | |
XM_006723854.3 | XP_006723917.1 | 11 - 12 | 1054 | 86 | |
XM_017028009.1 | XP_016883498.1 | 11 - 12 | 1426 | 86 | |
XM_017028010.1 | XP_016883499.1 | 10 - 11 | 929 | 86 | |
XM_017028011.1 | XP_016883500.1 | 10 - 11 | 1382 | 86 | |
GenBank mRNA | AB058737.1 | - | 11 - 12 | 1066 | 86 |
AF398886.1 | - | 10 - 11 | 994 | 86 | |
AF417861.1 | - | 10 - 11 | 926 | 86 | |
AK057032.1 | - | 12 - 13 | 1079 | 86 | |
AK293861.1 | - | 10 - 11 | 1019 | 86 | |
AK299318.1 | - | 14 - 15 | 1166 | 86 | |
AK302710.1 | - | 2 - 3 | 358 | 86 | |
AL713765.1 | - | 1 - 2 | 100 | 86 | |
BC000143.4 | - | 11 - 12 | 1131 | 86 | |
BC021079.2 | - | 1 - 2 | 118 | 86 | |
BX648819.1 | - | 11 - 12 | 1032 | 86 |