The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198783.1 | NP_001185712.1 | 2 - 3 | 330 | 90 |
NM_001198786.1 | NP_001185715.1 | 2 - 3 | 197 | 90 | |
NM_002697.3 | NP_002688.3 | 2 - 3 | 197 | 90 | |
NR_037163.1 | 3 - 4 | 241 | 90 | ||
XM_011509653.2 | XP_011507955.1 | 3 - 4 | 149 | 90 | |
XM_011509654.2 | XP_011507956.1 | 2 - 3 | 110 | 90 | |
XM_011509655.1 | XP_011507957.1 | 3 - 4 | 149 | 90 | |
XM_017001508.1 | XP_016856997.1 | 3 - 4 | 149 | 90 | |
GenBank mRNA | AK302525.1 | - | 3 - 4 | 236 | 90 |
AK303201.1 | - | 2 - 3 | 135 | 90 | |
AK308004.1 | - | 3 - 4 | 236 | 90 | |
AY113189.1 | - | 2 - 3 | 149 | 90 | |
BC001664.2 | - | 2 - 3 | 113 | 90 | |
BC003571.1 | - | 2 - 3 | 113 | 90 | |
BC052274.1 | - | 3 - 4 | 221 | 90 | |
BX954888.1 | - | 2 - 3 | 198 | 90 | |
DA937884.1 | - | 2 - 3 | 330 | 90 | |
DC370634.1 | - | 3 - 4 | 241 | 90 | |
KT438684.1 | - | 4 - 5 | 308 | 90 | |
X13403.1 | - | 2 - 3 | 118 | 90 |