Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POU2F1
Entrez Gene ID
5451
Gene Name
POU class 2 homeobox 1
Gene Aliases
OCT1, OTF1, oct-1B
Chromosome Location
Chr.1: 167220829 - 167427345 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198783.1 NP_001185712.1 3 - 4 426 65
NM_001198786.1 NP_001185715.1 3 - 4 293 65
NM_002697.3 NP_002688.3 3 - 4 293 65
NR_037163.1 4 - 5 337 65
XM_011509653.2 XP_011507955.1 4 - 5 245 65
XM_011509654.2 XP_011507956.1 3 - 4 206 65
XM_011509655.1 XP_011507957.1 4 - 5 245 65
XM_017001508.1 XP_016856997.1 4 - 5 245 65
GenBank mRNA AK091438.1 - 5 - 6 721 65
AK302525.1 - 4 - 5 332 65
AK303201.1 - 3 - 4 231 65
AK308004.1 - 4 - 5 332 65
AY113189.1 - 3 - 4 245 65
BC001664.2 - 3 - 4 209 65
BC003571.1 - 3 - 4 209 65
BC052274.1 - 4 - 5 317 65
DA937884.1 - 3 - 4 426 65
DC370634.1 - 4 - 5 337 65
KT438684.1 - 5 - 6 404 65
X13403.1 - 3 - 4 214 65

View Full Product Details