The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015336.2 | NP_056151.2 | 5 - 6 | 701 | 78 |
XM_005268750.4 | XP_005268807.1 | 5 - 6 | 541 | 78 | |
XM_011538083.1 | XP_011536385.1 | 5 - 6 | 565 | 78 | |
XM_017019091.1 | XP_016874580.1 | 5 - 6 | 541 | 78 | |
XM_017019092.1 | XP_016874581.1 | 4 - 5 | 445 | 78 | |
XM_017019093.1 | XP_016874582.1 | 4 - 5 | 394 | 78 | |
XM_017019094.1 | XP_016874583.1 | 4 - 5 | 422 | 78 | |
GenBank mRNA | AB023163.1 | - | 5 - 6 | 645 | 78 |
AB097013.1 | - | 5 - 6 | 521 | 78 | |
AB097035.1 | - | 5 - 6 | 521 | 78 | |
AK292866.1 | - | 5 - 6 | 517 | 78 | |
AK299089.1 | - | 4 - 5 | 413 | 78 | |
BC030990.1 | - | 4 - 5 | 568 | 78 | |
BC050324.1 | - | 5 - 6 | 660 | 78 |