Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPP1R12A
Entrez Gene ID
4659
Gene Name
protein phosphatase 1 regulatory subunit 12A
Gene Aliases
M130, MBS, MYPT1
Chromosome Location
Chr.12: 79773563 - 79935455 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143885.1 NP_001137357.1 9 - 10 1240 90
NM_001143886.1 NP_001137358.1 8 - 9 995 90
NM_001244990.1 NP_001231919.1 9 - 10 1240 90
NM_001244992.1 NP_001231921.1 8 - 9 1137 90
NM_002480.2 NP_002471.1 8 - 9 1384 90
XM_005268887.3 XP_005268944.1 8 - 9 1383 90
XM_011538374.1 XP_011536676.1 8 - 9 1373 90
XM_011538375.1 XP_011536677.1 8 - 9 1373 90
XM_011538376.2 XP_011536678.1 8 - 9 1373 90
XM_011538377.1 XP_011536679.1 7 - 8 1298 90
XM_011538378.2 XP_011536680.1 8 - 9 1373 90
XM_011538379.1 XP_011536681.1 8 - 9 1373 90
XM_011538380.1 XP_011536682.1 8 - 9 1373 90
XM_011538383.2 XP_011536685.1 8 - 9 1373 90
XM_011538384.1 XP_011536686.1 8 - 9 1373 90
XM_011538385.1 XP_011536687.1 8 - 9 1373 90
XM_017019323.1 XP_016874812.1 8 - 9 1373 90
XM_017019324.1 XP_016874813.1 7 - 8 1298 90
XM_017019325.1 XP_016874814.1 7 - 8 1298 90
XM_017019326.1 XP_016874815.1 8 - 9 1373 90
XM_017019327.1 XP_016874816.1 8 - 9 1373 90
XM_017019328.1 XP_016874817.1 7 - 8 1298 90
XM_017019329.1 XP_016874818.1 7 - 8 1298 90
XM_017019330.1 XP_016874819.1 8 - 9 1373 90
GenBank mRNA AF458589.1 - 9 - 10 1240 90
AK302692.1 - 8 - 9 995 90
AK314193.1 - 8 - 9 1124 90
AY380574.1 - 8 - 9 1117 90
BC047898.1 - 8 - 9 1307 90
BC062560.1 - 8 - 9 1116 90
BC092481.1 - 9 - 10 1240 90
BC111752.1 - 8 - 9 1117 90
D87930.1 - 8 - 9 1117 90

View Full Product Details