Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COLQ
Entrez Gene ID
8292
Gene Name
collagen like tail subunit of asymmetric acetylcholinesterase
Gene Aliases
CMS5, EAD
Chromosome Location
Chr.3: 15450133 - 15521751 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005677.3 NP_005668.2 13 - 14 1083 125
NM_080538.2 NP_536799.1 13 - 14 1212 125
NM_080539.3 NP_536800.2 12 - 13 981 125
GenBank mRNA AF057036.1 - 13 - 14 1083 125
AJ225895.1 - 13 - 14 1076 125
AK128401.1 - 13 - 14 1212 125
AK289688.1 - 13 - 14 1102 125
AY150334.1 - 13 - 14 1060 125
AY150335.1 - 13 - 14 1060 125
AY150336.1 - 13 - 14 1060 125
AY150337.1 - 13 - 14 1060 125
AY150338.1 - 13 - 14 1060 125
AY150339.1 - 13 - 14 1060 125
BC074828.2 - 13 - 14 987 125
BC074829.2 - 13 - 14 987 125

View Full Product Details