Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC91
Entrez Gene ID
55297
Gene Name
coiled-coil domain containing 91
Gene Aliases
HSD8, p56
Chromosome Location
Chr.12: 28190427 - 28550166 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018318.3 NP_060788.3 4 - 5 483 147
XM_005253413.1 XP_005253470.1 5 - 6 696 147
XM_005253414.1 XP_005253471.1 5 - 6 556 147
XM_005253415.1 XP_005253472.1 4 - 5 625 147
XM_006719103.3 XP_006719166.1 4 - 5 647 147
XM_006719104.2 XP_006719167.1 8 - 9 878 147
XM_006719105.1 XP_006719168.1 6 - 7 731 147
XM_011520729.2 XP_011519031.1 6 - 7 742 147
XM_011520731.2 XP_011519033.1 4 - 5 740 147
XM_011520733.2 XP_011519035.1 4 - 5 1067 147
XM_011520734.1 XP_011519036.1 3 - 4 544 147
XM_011520735.1 XP_011519037.1 3 - 4 499 147
XM_017019569.1 XP_016875058.1 7 - 8 771 147
XM_017019570.1 XP_016875059.1 5 - 6 665 147
XM_017019571.1 XP_016875060.1 6 - 7 702 147
XM_017019572.1 XP_016875061.1 7 - 8 665 147
XM_017019573.1 XP_016875062.1 4 - 5 768 147
XM_017019574.1 XP_016875063.1 6 - 7 662 147
XM_017019575.1 XP_016875064.1 8 - 9 862 147
XM_017019576.1 XP_016875065.1 8 - 9 878 147
XM_017019577.1 XP_016875066.1 5 - 6 696 147
GenBank mRNA AK093152.1 - 7 - 8 750 147
AK309181.1 - 5 - 6 556 147
AY289196.1 - 4 - 5 483 147
BC018679.1 - 4 - 5 901 147
BC028682.2 - 4 - 5 559 147
CR749586.1 - 1 - 2 146 147

View Full Product Details