The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001251845.1 | NP_001238774.1 | 5 - 6 | 898 | 105 |
NM_003304.4 | NP_003295.1 | 4 - 5 | 796 | 105 | |
XM_005247738.3 | XP_005247795.1 | 4 - 5 | 573 | 105 | |
XM_005247739.2 | XP_005247796.1 | 3 - 4 | 524 | 105 | |
XM_017007121.1 | XP_016862610.1 | 5 - 6 | 1506 | 105 | |
XM_017007122.1 | XP_016862611.1 | 3 - 4 | 1421 | 105 | |
GenBank mRNA | AB255425.1 | - | 4 - 5 | 660 | 105 |
BC112338.1 | - | 5 - 6 | 715 | 105 | |
BC113953.1 | - | 5 - 6 | 713 | 105 | |
X89066.1 | - | 4 - 5 | 797 | 105 |