The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146326.1 | NP_001139798.1 | 9 - 10 | 938 | 103 |
NM_001146327.1 | NP_001139799.1 | 9 - 10 | 1025 | 103 | |
NM_015363.4 | NP_056178.3 | 8 - 9 | 862 | 103 | |
GenBank mRNA | AF166122.1 | - | 8 - 9 | 845 | 103 |
BC104876.1 | - | 8 - 9 | 769 | 103 | |
BC104878.1 | - | 8 - 9 | 853 | 103 |