Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPP6R1
Entrez Gene ID
22870
Gene Name
protein phosphatase 6 regulatory subunit 1
Gene Aliases
KIAA1115, PP6R1, SAP190, SAPS1
Chromosome Location
Chr.19: 55229779 - 55258670 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014931.3 NP_055746.3 2 - 3 469 67
GenBank mRNA AB029038.2 - 2 - 3 794 67
AK297152.1 - 2 - 3 328 67
BC002799.2 - 1 - 2 71 67
BC068014.1 - 1 - 2 133 67
BC094753.1 - 2 - 3 331 67
BG772019.1 - 2 - 3 336 67
BX362839.2 - 1 - 2 169 67

View Full Product Details