Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCML2
Entrez Gene ID
10389
Gene Name
sex comb on midleg-like 2 (Drosophila)
Gene Aliases
-
Chromosome Location
Chr.X: 18239313 - 18355089 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006089.2 NP_006080.1 13 - 14 1979 79
NR_033717.1 13 - 14 1979 79
XM_017029219.1 XP_016884708.1 13 - 14 1875 79
XM_017029220.1 XP_016884709.1 13 - 14 1871 79
XM_017029222.1 XP_016884711.1 7 - 8 1257 79
GenBank mRNA AL833937.1 - 1 - 2 264 79
BC051913.1 - 13 - 14 1921 79
BC064617.1 - 13 - 14 1936 79
Y18004.1 - 13 - 14 1913 79

View Full Product Details