The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_147130.2 | NP_667341.1 | 3 - 4 | 764 | 107 |
XM_006715049.3 | XP_006715112.1 | 4 - 5 | 710 | 107 | |
XM_011514459.2 | XP_011512761.1 | 4 - 5 | 728 | 107 | |
GenBank mRNA | AF031137.1 | - | 3 - 4 | 764 | 107 |
BC052582.1 | - | 3 - 4 | 708 | 107 |